siRNA Details
| Gene Symbol: ZNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008727.3 | NP_001008727.1 | 4 | 1119 | ||
| NM_001308269.1 | NP_001295198.1 | 5 | 1177 | |||
| XM_017027239.1 | XP_016882728.1 | 5 | 1293 | |||
| GenBank mRNA | AK297919.1 | - | 5 | 1176 | ||
| AW949690.1 | - | 1 | 229 | |||

ZNF121
Genomic Map