siRNA Details
| Gene Symbol: Ubxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013093.1 | NP_001013111.1 | 2 | 360 | ||
| GenBank mRNA | BC079223.1 | - | 2 | 360 | 7132226 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubxn10| Gene Symbol: Ubxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013093.1 | NP_001013111.1 | 2 | 360 | ||
| GenBank mRNA | BC079223.1 | - | 2 | 360 | 7132226 | |