siRNA Details
| Gene Symbol: Igsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271454.1 | NP_001258383.1 | 7 | 1618 | ||
| XM_008764231.1 | XP_008762453.1 | 6 | 932 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf21| Gene Symbol: Igsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271454.1 | NP_001258383.1 | 7 | 1618 | ||
| XM_008764231.1 | XP_008762453.1 | 6 | 932 | |||