siRNA Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191766.1 | NP_001178695.1 | 15 | 1784 | ||
| XM_008764232.1 | XP_008762454.1 | 15 | 1852 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi6| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191766.1 | NP_001178695.1 | 15 | 1784 | ||
| XM_008764232.1 | XP_008762454.1 | 15 | 1852 | |||