siRNA Details
| Gene Symbol: Tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239327.2 | XP_006239389.1 | 5 | 1538 | ||
| XM_008764305.1 | XP_008762527.1 | 5 | 1679 | |||
| XM_008764306.1 | XP_008762528.1 | 5 | 1679 | |||
| XM_238429.8 | XP_238429.5 | 5 | 1538 | |||

Tmem82
Genomic Map