siRNA Details
| Gene Symbol: Slc25a34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013936.1 | NP_001013958.1 | 1 | 247 | ||
| GenBank mRNA | BC083723.1 | - | 1 | 247 | 7191003 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a34| Gene Symbol: Slc25a34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013936.1 | NP_001013958.1 | 1 | 247 | ||
| GenBank mRNA | BC083723.1 | - | 1 | 247 | 7191003 | |