siRNA Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048185.1 | NP_001041650.1 | 1 | 127 | ||
| XM_008764233.1 | XP_008762455.1 | 1 | 319 | |||
| GenBank mRNA | BC091231.1 | - | 1 | 111 | ||
| BC105628.1 | - | 1 | 127 | 7322772 | ||

Agmat
Genomic Map