siRNA Details
| Gene Symbol: Efhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031648.1 | NP_001026818.1 | 1, 2 | 324 | ||
| GenBank mRNA | BC098936.1 | - | 1, 2 | 324 | 7446346 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efhd2| Gene Symbol: Efhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031648.1 | NP_001026818.1 | 1, 2 | 324 | ||
| GenBank mRNA | BC098936.1 | - | 1, 2 | 324 | 7446346 | |