siRNA Details
Gene Symbol: Pramef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238356.2 | XP_006238418.1 | 3 | 1316 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pramef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238356.2 | XP_006238418.1 | 3 | 1316 |