siRNA Details
| Gene Symbol: Miip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017450.1 | NP_001017450.1 | 5 | 800 | ||
| XM_006239379.2 | XP_006239441.1 | 5 | 875 | |||
| XM_006239380.2 | XP_006239442.1 | 5 | 973 | |||
| XM_006239381.2 | XP_006239443.1 | 5 | 902 | |||
| GenBank mRNA | BC079047.1 | - | 5 | 800 | 7115212 | |
| FJ618907.1 | - | 5 | 1004 | |||
| FQ227320.1 | - | 5 | 758 | |||
| Gene Symbol: LOC100911456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239406.2 | XP_006239468.1 | 875 | |||
| XM_006239407.2 | XP_006239469.1 | 973 | ||||
| XM_006239408.2 | XP_006239470.1 | 999 | ||||
| XM_006239409.2 | XP_006239471.1 | 902 | ||||

Miip
Genomic Map