siRNA Details
Gene Symbol: Tardbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011979.2 | NP_001011979.1 | 4 | 562 | ||
XM_006239382.2 | XP_006239444.1 | 4 | 615 | |||
XM_006239384.2 | XP_006239446.1 | 4 | 592 | |||
XM_006239385.2 | XP_006239447.1 | 4 | 593 | |||
XM_006239386.2 | XP_006239448.1 | 4 | 597 | |||
XM_008764234.1 | XP_008762456.1 | 4 | 594 | |||
XM_008764235.1 | XP_008762457.1 | 4 | 592 | |||
XM_008764236.1 | XP_008762458.1 | 4 | 592 | |||
GenBank mRNA | BC083752.1 | - | 4 | 551 | 7192591 |
