siRNA Details
| Gene Symbol: Tardbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011979.2 | NP_001011979.1 | 2 | 324 | ||
| XM_006239382.2 | XP_006239444.1 | 2 | 377 | |||
| XM_006239384.2 | XP_006239446.1 | 2 | 354 | |||
| XM_006239385.2 | XP_006239447.1 | 2 | 355 | |||
| XM_006239386.2 | XP_006239448.1 | 2 | 359 | |||
| XM_008764234.1 | XP_008762456.1 | 2 | 356 | |||
| XM_008764235.1 | XP_008762457.1 | 2 | 354 | |||
| XM_008764236.1 | XP_008762458.1 | 2 | 354 | |||
| GenBank mRNA | AI502405.1 | - | 2 | 335 | ||
| BC083752.1 | - | 2 | 313 | 7192591 | ||

Tardbp
Genomic Map