siRNA Details
| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037556.2 | NP_001032645.2 | 3 | 291 | ||
| XM_006239429.2 | XP_006239491.1 | 3 | 925 | |||
| XM_006239430.2 | XP_006239492.1 | 3 | 510 | |||
| XM_006239431.2 | XP_006239493.1 | 2 | 252 | |||
| XM_008764237.1 | XP_008762459.1 | 4 | 448 | |||
| XM_008764238.1 | XP_008762460.1 | 3 | 273 | |||
| GenBank mRNA | AY788917.1 | - | 3 | 285 | ||
| BC127446.1 | - | 3 | 291 | 7383226 | ||

Nmnat1
Genomic Map