siRNA Details
| Gene Symbol: Rer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039012.1 | NP_001034101.1 | 5 | 355 | ||
| GenBank mRNA | BC100270.1 | - | 5 | 355 | 7444634 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rer1| Gene Symbol: Rer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039012.1 | NP_001034101.1 | 5 | 355 | ||
| GenBank mRNA | BC100270.1 | - | 5 | 355 | 7444634 | |