siRNA Details
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013938.1 | NP_001013960.1 | 3 | 940 | ||
| GenBank mRNA | BC081983.1 | - | 3 | 940 | 7128587 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa1| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013938.1 | NP_001013960.1 | 3 | 940 | ||
| GenBank mRNA | BC081983.1 | - | 3 | 940 | 7128587 | |