siRNA Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013094.1 | NP_001013112.1 | 4 | 523 | ||
| XM_006239544.2 | XP_006239606.1 | 4 | 1045 | |||
| XM_006239545.2 | XP_006239607.1 | 4 | 1044 | |||
| GenBank mRNA | BC088316.1 | - | 4 | 523 | 7325177 | |
| FQ211043.1 | - | 4 | 584 | |||

Ccnl2
Genomic Map