siRNA Details
| Gene Symbol: Cpsf3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033892.1 | NP_001029064.1 | 9 | 1021 | ||
| XM_006239549.2 | XP_006239611.1 | 9 | 1063 | |||
| XM_006239550.2 | XP_006239612.1 | 9 | 1063 | |||
| XM_006239551.2 | XP_006239613.1 | 9 | 1063 | |||
| XM_006239552.2 | XP_006239614.1 | 9 | 1091 | |||
| XM_006239553.2 | XP_006239615.1 | 7 | 920 | |||
| GenBank mRNA | BC105303.1 | - | 9 | 1021 | 7110348 | |

Cpsf3l
Genomic Map