siRNA Details
| Gene Symbol: Ndufaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008318.1 | NP_001008319.1 | 10 | 1174 | ||
| XM_006239627.2 | XP_006239689.1 | 10 | 1281 | |||
| GenBank mRNA | BC083810.1 | - | 10 | 1174 | 7110144 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufaf7| Gene Symbol: Ndufaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008318.1 | NP_001008319.1 | 10 | 1174 | ||
| XM_006239627.2 | XP_006239689.1 | 10 | 1281 | |||
| GenBank mRNA | BC083810.1 | - | 10 | 1174 | 7110144 | |