siRNA Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277279.1 | NP_001264208.1 | 6 | 393 | ||
| GenBank mRNA | BC167086.1 | - | 5 | 360 | 5621310 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a6| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277279.1 | NP_001264208.1 | 6 | 393 | ||
| GenBank mRNA | BC167086.1 | - | 5 | 360 | 5621310 | |