siRNA Details
| Gene Symbol: ZNF136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003437.3 | NP_003428.1 | 3 | 308 | ||
| XM_011528269.2 | XP_011526571.1 | 4 | 803 | |||
| XM_017027243.1 | XP_016882732.1 | 2 | 566 | |||
| GenBank mRNA | AK123008.1 | - | 2 | 1325 | ||
| AK291242.1 | - | 3 | 303 | |||
| AK299601.1 | - | 2 | 131 | |||
| BC006421.1 | - | 3 | 261 | 4040430 | ||
| DB200139.1 | - | 3 | 308 | |||
| U09367.1 | - | 3 | 232 | |||

ZNF136
Genomic Map