siRNA Details
| Gene Symbol: LOC298795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013941.1 | NP_001013963.1 | 1 | 559 | ||
| GenBank mRNA | BC079389.1 | - | 1 | 548 | ||
| BC089860.1 | - | 1 | 559 | 7112967 | ||
| Gene Symbol: LOC103690862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773315.1 | XP_008771537.1 | 2 | 195 | ||
| Gene Symbol: Sfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750048.3 | XP_003750096.1 | 1 | 829 | ||

LOC298795
Genomic Map