siRNA Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013942.1 | NP_001013964.1 | 10, 11 | 1414 | ||
| GenBank mRNA | BC081910.1 | - | 10, 11 | 1414 | 7111548 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clip4| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013942.1 | NP_001013964.1 | 10, 11 | 1414 | ||
| GenBank mRNA | BC081910.1 | - | 10, 11 | 1414 | 7111548 | |