siRNA Details
Gene Symbol: Clip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013942.1 | NP_001013964.1 | 5 | 573 | ||
GenBank mRNA | BC081910.1 | - | 5 | 573 | 7111548 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013942.1 | NP_001013964.1 | 5 | 573 | ||
GenBank mRNA | BC081910.1 | - | 5 | 573 | 7111548 |