siRNA Details
| Gene Symbol: ZNF136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003437.3 | NP_003428.1 | 4 | 1605 | ||
| XM_011528269.2 | XP_011526571.1 | 5 | 2100 | |||
| XM_017027243.1 | XP_016882732.1 | 3 | 1863 | |||
| GenBank mRNA | AK123008.1 | - | 3 | 2622 | ||
| AK291242.1 | - | 4 | 1600 | |||
| AK299601.1 | - | 3 | 1428 | |||
| AY500356.1 | - | 1 | 120 | |||
| BC006421.1 | - | 4 | 1558 | 4040430 | ||
| U09367.1 | - | 4 | 1529 | |||

ZNF136
Genomic Map