siRNA Details
| Gene Symbol: Snx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011981.1 | NP_001011981.1 | 6 | 576 | ||
| GenBank mRNA | BC078931.1 | - | 6 | 576 | 7107232 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx17| Gene Symbol: Snx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011981.1 | NP_001011981.1 | 6 | 576 | ||
| GenBank mRNA | BC078931.1 | - | 6 | 576 | 7107232 | |