siRNA Details
| Gene Symbol: Slc35f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017451.1 | NP_001017451.1 | 908 | |||
| GenBank mRNA | BC085904.1 | - | 908 | 7193745 | ||
| FQ213959.1 | - | 1335 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35f6| Gene Symbol: Slc35f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017451.1 | NP_001017451.1 | 908 | |||
| GenBank mRNA | BC085904.1 | - | 908 | 7193745 | ||
| FQ213959.1 | - | 1335 | ||||