siRNA Details
| Gene Symbol: Gpr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106712.1 | NP_001100182.1 | 9 | 2768 | ||
| XM_006239800.2 | XP_006239862.1 | 10 | 3041 | |||
| XM_006239801.2 | XP_006239863.1 | 9 | 2900 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr113| Gene Symbol: Gpr113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106712.1 | NP_001100182.1 | 9 | 2768 | ||
| XM_006239800.2 | XP_006239862.1 | 10 | 3041 | |||
| XM_006239801.2 | XP_006239863.1 | 9 | 2900 | |||