siRNA Details
| Gene Symbol: ZNF137P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023311.1 | 1 | 180 | |||
| GenBank mRNA | BC110455.1 | - | 1 | 180 | ||
| BC110456.1 | - | 1 | 180 | |||
| U09414.1 | - | 1 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF137P| Gene Symbol: ZNF137P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023311.1 | 1 | 180 | |||
| GenBank mRNA | BC110455.1 | - | 1 | 180 | ||
| BC110456.1 | - | 1 | 180 | |||
| U09414.1 | - | 1 | 214 | |||