siRNA Details
| Gene Symbol: Pum2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106715.1 | NP_001100185.1 | 15 | 2335 | ||
| XM_006239879.2 | XP_006239941.1 | 15 | 2425 | |||
| XM_006239880.2 | XP_006239942.1 | 15 | 2425 | |||
| XM_006239883.2 | XP_006239945.1 | 14 | 2188 | |||
| XM_006239884.2 | XP_006239946.2 | 14 | 3101 | |||
| XM_008764579.1 | XP_008762801.1 | 14 | 3100 | |||
| XM_008764580.1 | XP_008762802.1 | 15 | 2446 | |||
| XM_008764581.1 | XP_008762803.1 | 13 | 2863 | |||
| XM_008764582.1 | XP_008762804.1 | 13 | 2862 | |||
| GenBank mRNA | AY724506.1 | - | 13 | 1953 | ||
| FQ232361.1 | - | 3 | 493 | |||

Pum2
Genomic Map