siRNA Details
| Gene Symbol: Laptm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199384.2 | NP_955416.1 | 4 | 446 | ||
| GenBank mRNA | BC063179.1 | - | 4 | 450 | 6890564 | |
| FQ212732.1 | - | 4 | 848 | |||
| FQ213363.1 | - | 4 | 480 | |||
| FQ229677.1 | - | 405 | ||||
| FQ232171.1 | - | 4 | 504 | |||
| FQ232930.1 | - | 4 | 464 | |||
| FQ235113.1 | - | 4 | 351 | |||

Laptm4a
Genomic Map