siRNA Details
| Gene Symbol: Fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106718.2 | NP_001100188.1 | 5 | 466 | ||
| XM_008764584.1 | XP_008762806.1 | 5 | 1015 | |||
| XM_008764585.1 | XP_008762807.1 | 4 | 368 | |||
| GenBank mRNA | BC158700.1 | - | 5 | 466 | 9028479 | |

Fam49a
Genomic Map