siRNA Details
| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034952.1 | NP_001030124.1 | 2 | 329 | ||
| XM_008764587.1 | XP_008762809.1 | 2 | 652 | |||
| GenBank mRNA | BC081879.1 | - | 2 | 235 | 7108131 | |
| CK477351.1 | - | 2 | 152 | |||

Pqlc3
Genomic Map