siRNA Details
| Gene Symbol: Kcnf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169104.1 | NP_001162575.1 | 1 | 608 | ||
| GenBank mRNA | M81783.1 | - | 1 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnf1| Gene Symbol: Kcnf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169104.1 | NP_001162575.1 | 1 | 608 | ||
| GenBank mRNA | M81783.1 | - | 1 | 614 | ||