siRNA Details
| Gene Symbol: Itgb1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106719.2 | NP_001100189.1 | 5, 6 | 573 | ||
| XM_006239979.2 | XP_006240041.1 | 4, 5 | 489 | |||
| XM_006239980.2 | XP_006240042.1 | 5, 6 | 518 | |||
| XM_006239981.2 | XP_006240043.1 | 4, 5 | 700 | |||
| GenBank mRNA | BC169104.1 | - | 5, 6 | 573 | 7386477 | |
| FQ215419.1 | - | 4, 5 | 408 | |||
| FQ218542.1 | - | 4, 5 | 450 | |||
| FQ229091.1 | - | 5, 6 | 576 | |||

Itgb1bp1
Genomic Map