siRNA Details
| Gene Symbol: Iah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100540.1 | NP_001094010.1 | 4 | 353 | ||
| GenBank mRNA | BC168771.1 | - | 4 | 353 | 6919650 | |
| FQ220941.1 | - | 4 | 349 | |||
| FQ225724.1 | - | 4 | 378 | |||
| FQ231316.1 | - | 4 | 378 | |||
| FQ232084.1 | - | 4 | 383 | |||
| FQ235324.1 | - | 4 | 378 | |||

Iah1
Genomic Map