siRNA Details
| Gene Symbol: Rnaseh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013097.2 | NP_001013115.1 | 7 | 774 | ||
| NM_001286938.1 | NP_001273867.1 | 7 | 774 | |||
| GenBank mRNA | BC091209.1 | - | 7 | 749 | 7381386 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnaseh1| Gene Symbol: Rnaseh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013097.2 | NP_001013115.1 | 7 | 774 | ||
| NM_001286938.1 | NP_001273867.1 | 7 | 774 | |||
| GenBank mRNA | BC091209.1 | - | 7 | 749 | 7381386 | |