siRNA Details
| Gene Symbol: Adi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199097.1 | NP_954528.1 | 1, 2 | 143 | ||
| GenBank mRNA | AY346335.1 | - | 1, 2 | 143 | ||
| BC092562.1 | - | 1, 2 | 132 | 7326245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adi1| Gene Symbol: Adi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199097.1 | NP_954528.1 | 1, 2 | 143 | ||
| GenBank mRNA | AY346335.1 | - | 1, 2 | 143 | ||
| BC092562.1 | - | 1, 2 | 132 | 7326245 | ||