siRNA Details
| Gene Symbol: Sntg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106720.1 | NP_001100190.1 | 6 | 696 | ||
| XM_006239967.2 | XP_006240029.1 | 10 | 977 | |||
| XM_006239968.2 | XP_006240030.1 | 10 | 977 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sntg2| Gene Symbol: Sntg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106720.1 | NP_001100190.1 | 6 | 696 | ||
| XM_006239967.2 | XP_006240029.1 | 10 | 977 | |||
| XM_006239968.2 | XP_006240030.1 | 10 | 977 | |||