siRNA Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106726.1 | NP_001100196.1 | 6 | 475 | ||
| XM_006240078.2 | XP_006240140.1 | 5 | 401 | |||
| XM_006240079.2 | XP_006240141.1 | 7 | 566 | |||
| XM_006240080.2 | XP_006240142.1 | 6 | 1928 | |||
| GenBank mRNA | FQ221669.1 | - | 7 | 1396 | ||

G2e3
Genomic Map