siRNA Details
| Gene Symbol: Eif2s3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100542.1 | NP_001094012.1 | 9 | 990 | ||
| GenBank mRNA | BC091286.1 | - | 7 | 773 | ||
| BC166783.1 | - | 9 | 972 | 7935021 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2s3x| Gene Symbol: Eif2s3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100542.1 | NP_001094012.1 | 9 | 990 | ||
| GenBank mRNA | BC091286.1 | - | 7 | 773 | ||
| BC166783.1 | - | 9 | 972 | 7935021 | ||