siRNA Details
| Gene Symbol: RGD1305089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106730.2 | NP_001100200.1 | 1 | 862 | ||
| GenBank mRNA | BC168878.1 | - | 1 | 862 | 7382020 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305089| Gene Symbol: RGD1305089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106730.2 | NP_001100200.1 | 1 | 862 | ||
| GenBank mRNA | BC168878.1 | - | 1 | 862 | 7382020 | |