siRNA Details
| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106733.1 | NP_001100203.1 | 3 | 404 | ||
| GenBank mRNA | FQ230024.1 | - | 3 | 167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc6b| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106733.1 | NP_001100203.1 | 3 | 404 | ||
| GenBank mRNA | FQ230024.1 | - | 3 | 167 | ||