siRNA Details
| Gene Symbol: PCGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007144.2 | NP_009075.1 | 9 | 802 | ||
| XM_005257640.2 | XP_005257697.1 | 10 | 1261 | |||
| XM_005257641.4 | XP_005257698.1 | 8 | 1483 | |||
| XM_005257642.3 | XP_005257699.1 | 8 | 1880 | |||
| XM_017025016.1 | XP_016880505.1 | 10 | 1821 | |||
| GenBank mRNA | BC004858.2 | - | 7 | 573 | 3841545 | |
| BC024255.1 | - | 9 | 798 | 3605568 | ||
| BX647429.1 | - | 10 | 1261 | |||
| D13969.1 | - | 8 | 760 | |||

PCGF2
Genomic Map