siRNA Details
| Gene Symbol: Dhrs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013098.1 | NP_001013116.1 | 5 | 816 | ||
| GenBank mRNA | AB108671.1 | - | 5 | 816 | ||
| BC104713.1 | - | 5 | 722 | 7370181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs7| Gene Symbol: Dhrs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013098.1 | NP_001013116.1 | 5 | 816 | ||
| GenBank mRNA | AB108671.1 | - | 5 | 816 | ||
| BC104713.1 | - | 5 | 722 | 7370181 | ||