siRNA Details
| Gene Symbol: Tex21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025658.1 | NP_001020829.1 | 2 | 590 | ||
| XM_006240226.2 | XP_006240288.1 | 4 | 793 | |||
| XM_006240227.2 | XP_006240289.1 | 4 | 837 | |||
| XM_008764743.1 | XP_008762965.1 | 4 | 837 | |||
| XM_008764744.1 | XP_008762966.1 | 3 | 708 | |||
| XM_008764745.1 | XP_008762967.1 | 2 | 313 | |||
| GenBank mRNA | BC097422.1 | - | 2 | 590 | 7454996 | |

Tex21
Genomic Map