siRNA Details
| Gene Symbol: Ppp1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013944.1 | NP_001013966.1 | 9 | 791 | ||
| XM_006240231.1 | XP_006240293.1 | 9 | 880 | |||
| GenBank mRNA | BC079166.1 | - | 9 | 791 | 7129668 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r36| Gene Symbol: Ppp1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013944.1 | NP_001013966.1 | 9 | 791 | ||
| XM_006240231.1 | XP_006240293.1 | 9 | 880 | |||
| GenBank mRNA | BC079166.1 | - | 9 | 791 | 7129668 | |