siRNA Details
| Gene Symbol: Rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198749.2 | NP_942044.1 | 5 | 742 | ||
| GenBank mRNA | BC094954.1 | - | 5 | 742 | 7316003 | |
| M83679.1 | - | 5 | 586 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab15| Gene Symbol: Rab15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198749.2 | NP_942044.1 | 5 | 742 | ||
| GenBank mRNA | BC094954.1 | - | 5 | 742 | 7316003 | |
| M83679.1 | - | 5 | 586 | |||