siRNA Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015009.1 | NP_001015009.1 | 6 | 884 | ||
| XM_006240326.1 | XP_006240388.1 | 6 | 898 | |||
| XM_006240327.1 | XP_006240389.1 | 6 | 834 | |||
| XM_006240328.2 | XP_006240390.1 | 6 | 796 | |||
| XM_006240329.2 | XP_006240391.1 | 4 | 1421 | |||
| GenBank mRNA | BC091173.1 | - | 6 | 884 | 7189599 | |

Ptgr2
Genomic Map