siRNA Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011983.1 | NP_001011983.1 | 4 | 431 | ||
| GenBank mRNA | BC079342.1 | - | 4 | 431 | 7106851 | |
| FQ228932.1 | - | 3 | 788 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq6| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011983.1 | NP_001011983.1 | 4 | 431 | ||
| GenBank mRNA | BC079342.1 | - | 4 | 431 | 7106851 | |
| FQ228932.1 | - | 3 | 788 | |||