siRNA Details
| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013100.3 | NP_001013118.2 | 6, 7 | 720 | ||
| XM_006240330.2 | XP_006240392.1 | 6, 7 | 807 | |||
| XM_006240331.2 | XP_006240393.1 | 6, 7 | 814 | |||
| GenBank mRNA | CB785701.1 | - | 2, 3 | 234 | ||

Abcd4
Genomic Map